Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N7756

Sigma-Aldrich

N-Nitrosodimethylamine

Synonym(s):

Dimethylnitrosamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NNO
CAS Number:
Molecular Weight:
74.08
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

vapor pressure

5 mmHg ( 20 °C)

description

Bulk package

refractive index

n20/D 1.437 (lit.)

bp

153 °C/774 mmHg (lit.)

density

1.01 g/mL (lit.)

storage temp.

2-8°C

SMILES string

CN(C)N=O

InChI

1S/C2H6N2O/c1-4(2)3-5/h1-2H3

InChI key

UMFJAHHVKNCGLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Induces gastric, liver, kidney and lung cancer in mice and rats.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Carc. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yu Peng et al.
European journal of clinical investigation, 43(4), 358-368 (2013-02-27)
Fluorofenidone (AKF-PD) is a novel pyridone agent. The purpose of this study is to investigate the inhibitory effects of AKF-PD on dimethylnitrosamine (DMN)-induced liver fibrosis in rats and the involved molecular mechanism related to hepatic stellate cells (HSCs). Wistar rats
Man-Chun Huang et al.
Food chemistry, 138(1), 227-233 (2012-12-26)
A sensitive procedure, microwave-assisted extraction (MAE) coupled dispersive micro solid-phase extraction (D-μ-SPE), was developed to extract N-nitrosodimethylamine (NDMA) and other six volatile N-nitrosamines (NAms) from meat products. Parameters affecting the efficiency of MAE and D-μ-SPE were systematically investigated. For MAE
Maria José Farré et al.
Environmental science & technology, 46(23), 12904-12912 (2012-11-09)
UV-C radiation is the U.S. EPA recommended technology to remove N-nitrosodimethylamine (NDMA) during drinking and recycled water production. Frequently, H(2)O(2) is added to the treatment to remove other recalcitrant compounds and to prevent NDMA reformation. However, the transformation of NDMA
Xiaojie Gan et al.
Water research, 47(3), 1344-1352 (2013-01-05)
The objective of this study was to examine the potential of Magnetic Ion Exchange (MIEX(®)) technology for removing nitrogenous disinfection byproduct (N-DBP) precursors while minimizing carbonaceous DBP (C-DBP) precursors in (i) surface waters, and (ii) effluent impacted waters. Samples were
David Hanigan et al.
Environmental science & technology, 46(22), 12630-12639 (2012-10-31)
Activated carbon (AC) has been shown to remove precursors of halogenated disinfection byproducts. Granular and powdered activated carbon (GAC, PAC) were investigated for their potential to adsorb N-nitrosodimethylamine (NDMA) precursors from blends of river water and effluent from a wastewater

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service