MilliporeSigma
  • The antihepatic fibrotic effects of fluorofenidone via MAPK signalling pathways.
The antihepatic fibrotic effects of fluorofenidone via MAPK signalling pathways.
European journal of clinical investigation (2013-02-27)
Yu Peng, Huixiang Yang, Tingting Zhu, Menghua Zhao, Yuexia Deng, Bin Liu, Hong Shen, Gaoyun Hu, Zhaohe Wang, Lijian Tao
ABSTRACT
Fluorofenidone (AKF-PD) is a novel pyridone agent. The purpose of this study is to investigate the inhibitory effects of AKF-PD on dimethylnitrosamine (DMN)-induced liver fibrosis in rats and the involved molecular mechanism related to hepatic stellate cells (HSCs). Wistar rats were randomly divided into normal control, DMN, DMN/AKF-PD treatment and DMN/pirfenidone (PFD) treatment groups. AKF-PD and PFD treatments were, respectively, performed for two activated HSCs lines, rat CFSC-2G and human LX2. The cell proliferation was analysed by MTT. The expression of collagen I was determined by immunohistochemical staining and real-time RT-PCR. The expression of α-smooth muscle actin (α-SMA), tissue inhibitor of metalloproteinases-1 (TIMP-1), extracellular signal regulated kinase (ERK1/2), p38 MAPK (p38), and c-Jun N-terminal kinase/stress-activated protein kinase (JNK) were also detected by real-time RT-PCR and/or Western blot. AKF-PD significantly reduced PDGF-BB-induced proliferation and activation of HSCs, as determined by reducing protein expression of α-SMA and TIMP-1. AKF-PD treatment attenuated PDGF-BB-induced upregulation of phosphorylation of ERK1/2, p38 and JNK. In fibrotic rat liver, AKF-PD reduced the degree of liver injury and hepatic fibrosis, which was associated with reduced the expression of collagen I, α-SMA, TIMP-1 at both mRNA and protein levels. AKF-PD treatment inhibits the progression of hepatic fibrosis by suppressing HSCs proliferation and activation via MAPK signalling pathway.
Product Description
Supelco
N-Nitrosodimethylamine (NDMA) solution, certified reference material, 200 μg/mL in methanol