Skip to Content
MilliporeSigma
All Photos(1)

Documents

591068

Sigma-Aldrich

N-Nitrosodimethylamine-d6

98 atom % D

Synonym(s):

Deuterated N-nitrosodimethylamine, Dimethylnitrosamine-d6, NDMA-d6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2NNO
CAS Number:
Molecular Weight:
80.12
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Assay

98% (CP)

bp

153 °C/774 mmHg (lit.)

density

1.086 g/mL at 25 °C

mass shift

M+6

SMILES string

[2H]C([2H])([2H])N(N=O)C([2H])([2H])[2H]

InChI

1S/C2H6N2O/c1-4(2)3-5/h1-2H3/i1D3,2D3

InChI key

UMFJAHHVKNCGLG-WFGJKAKNSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Carc. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Dong et al.
Journal of chromatography. A, 1422, 82-88 (2015-11-01)
A sensitive gas chromatography-mass spectrometry method was established for the simultaneous determination of thirteen N-nitrosamines (NAs) in skin care cosmetics. The cosmetics samples were firstly dispersed by water and subsequently extracted and purified using salting out-acetonitrile homogeneous extraction method. Finally

Articles

An overview of nitrosamine impurity testing, worldwide regulations, and key considerations in filter selection for sample preparation and analysis.

This application note describes the LC-MS-based quantitative analysis of known nitrosamine impurities following procedures 1 and 3 as given in USP general chapter <1469>.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service