Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MTCT1411C

MTC Bio Plug Cap For Test Tubes

for use with 13mm, rimless tubes, polyethylene

Synonym(s):

MTC Bio Plug Cap For Test Tubes, plastic tube cap, reagent dish, tube cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122500
NACRES:
NB.11

material

polyethylene

sterility

non-sterile

packaging

bag of 20 × 1000 ea

manufacturer/tradename

MTC Bio T1411-C

Looking for similar products? Visit Product Comparison Guide

General description

Non-sterile plug caps for use with 13mm, rimless test tubes.

Legal Information

MTC is a trademark of MTC Bio

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service