CLS8422100
Pyrex® heavy-duty round-bottom centrifuge tubes, with phenolic screw cap and rubber liner
capacity 100 mL
Synonym(s):
Corning® 8422
Select a Size
$91.90
Select a Size
About This Item
$91.90
Recommended Products
material
borosilicate glass
cap (rubber liner)
round bottom tube
sterility
non-sterile
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 8422-100
availability
not available in EU
O.D. × L
43 mm × 139 mm
cap diam.
38 mm
cap thread
38
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Pyrex® heavy-duty round-bottom centrifuge tubes are made of low-expansion borosilicate glass.
- Best suited for digestion, shaking, neutralizing, and centrifuging applications in various clinical and microbiological tests.
- Features temperature and steam resistant screw caps for optimal product recovery.
- Includes black phenolic threaded cap (GPI 24-410) with a glued-in, white rubber liner.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service