Skip to Content
MilliporeSigma
All Photos(2)

Documents

MAK026

Sigma-Aldrich

Magnesium Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

application(s)

cosmetics
food and beverages

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; cardiovascular diseases

storage temp.

−20°C

General description

The new Magnesium Assay Kit, MAK507, is now available! Magnesium, one of the most abundant trace metals in living organisms, contributes to a variety of biological functions including ATP and nucleic acid processing, energy metabolism, and enzymatic function. Magnesium forms complexes with multiple molecules such as phospholipids and ATP. Low serum levels of magnesium have been associated with metabolic syndrome, diabetes mellitus type 2, and hypertension, as well as, increased smooth muscle and platelet reactivity.

Application

Magnesium Assay Kit has been used to quantify magnesium levels in conditioned media.

Suitability

Suitable for quantitating magnesium concentrations in a variety of biological samples.

Principle

The Magnesium Assay kit provides a simple and direct procedure for measuring magnesium in a variety of samples. The magnesium concentration is determined by a coupled enzyme assay that takes advantage of the specific requirement of glycerol kinase for Mg2+, resulting in a colorimetric (450 nm) product proportional to the magnesium present. This assay gives a linear range of 3-15 nmoles of magnesium and exhibits no detectable interference with Fe2+, Cu2+, Ni2+, Zn2+, Co2+, Ca2+, and Mn2+.

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnesium: its proven and potential clinical significance.
Fox C, et al.
Southern Medical Journal, 94(12), 1195-1201 (2001)
Microbial competition between Escherichia coli and Candida albicans reveals a soluble fungicidal factor.
Damien J C, et al.
Microbial cell, 5(5), 249-255 (2018)
Christine Moore Sheridan et al.
Malaria journal, 17(1), 465-465 (2018-12-14)
The continued spectre of resistance to existing anti-malarials necessitates the pursuit of novel targets and mechanisms of action for drug development. One class of promising targets consists of the 80S ribosome and its associated components comprising the parasite translational apparatus.
Damien J Cabral et al.
Microbial cell (Graz, Austria), 5(5), 249-255 (2018-05-26)
Localized and systemic fungal infections caused by Candida albicans can lead to significant mortality and morbidity. However, severe C. albicans infections are relatively rare, occurring mostly in the very young, the very old, and immunocompromised individuals. The fact that these
Ruth Edwige Kemadjou Dibacto et al.
Heliyon, 8(7), e09896-e09896 (2022-07-21)
Neurodegenerative disease such as Alzheimer's disease, are progressive disorders which has been linked to oxidative imbalance and associated perturbations characterised by loss of memory, cognition and cholinergic deficit. To date, cholinesterase inhibition and neuroprotection are the two major strategies in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service