Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.14815

Supelco

Calcium Test, photometric

photometric, 5-160 mg/L (Ca), 12-400 mg/L (CaO3), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Calcium Test, photometric, 5-160 mg/L (Ca), 12-400 mg/L (CaO3), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 100 tests
100 tests

specific analyte(s)

calcium

measuring range

12-400 mg/L (CaO3)
5-160 mg/L (Ca)
7-224 mg/L (CaO)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Glyoxalbis-hydroxyanil)

storage temp.

15-25°C

General description

Method: photometric 5 - 160 mg/l Ca 7 - 224 mg/l CaO 12 - 400 mg/l CaCO₃ Spectroquant®

This Spectroquant® Calcium Reagent Test allows the accurate quantification of the calcium content in aqueous samples.

Method applied: In alcoholic-alkaline solution calcium ions react with modified glyoxal-bis(2-hydroxyanil) to form a red-violet complex which is determined photometrically.

The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service