MilliporeSigma
  • The neuroprotective effect of Xylopia parviflora against aluminum chloride-induced neurotoxicity in rats.
The neuroprotective effect of Xylopia parviflora against aluminum chloride-induced neurotoxicity in rats.
Heliyon (2022-07-21)
Ruth Edwige Kemadjou Dibacto, Bruno Dupon Akamba Ambamba, Fils Armand Ella, Christine Fernande Biyegue Nyangono, Jules Vidal Kamga Nanhah, Martin Fonkoua, Réné Samuel Minka, Judith Laure Ngondi
ABSTRACT
Neurodegenerative disease such as Alzheimer's disease, are progressive disorders which has been linked to oxidative imbalance and associated perturbations characterised by loss of memory, cognition and cholinergic deficit. To date, cholinesterase inhibition and neuroprotection are the two major strategies in drug development. Xylopia parviflora (Annonacea family) is a spice consumed in Cameroon and has been used in traditional medicine to treat various pains. In this study, X. parviflora was evaluated on behavioural studies, ion homeostasis, cholinesterase inhibitory and antioxidant activities. Rats were exposed to aluminium chloride (75 mg/kg) during 60 days, and were treated with the extract of X. parviflora (150 and 300 mg/kg BW) and two drugs references (Donepezil and Curcumin). Behavioural parameters were assessed using the Morris-Maze test and the Open Field, followed by biochemical investigations, namely, cholinesterase enzyme activity (AChE and BChE), oxidative stress (NO, MDA, GSH level, SOD and Catalase activities) and ion homeostasis (Mg2+ and Ca2+ levels). AlCl3 administration shows a decrease in learning and memory improvement during behavioural studies, significant alteration of the central cholinergic system characterised by an increase in AChE and BChE activities to 2.72 ± 0.002 mol/min/g and 5.74 ± 0.12 mol/min/g respectively, disturbance of ion homeostasis with an increase in Ca2+ level (25.68 ± 3.78 μmol/mg protein) and a decrease in Mg2+ level (15.97 ± 2.05 μmol/mg protein) and an increase in oxidative stress compared to the positive control group. Treatment with the different doses of X. parviflora increased memory and improved locomotion, improved cholinesterase activities, ion homeostasis and stabilized brain oxidative stress levels. The study suggests that X. parviflora could potentially be used for the management of some biochemical alterations associated with Alzheimer's disease. It could even be a good alternative to chemical drugs for neurotoxicity and memory enhancement.