Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L8757

Sigma-Aldrich

Lysing Enzymes from Rhizoctonia solani

crude powder

Synonym(s):

Kitalase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

crude powder

storage temp.

2-8°C

Application

Lysing enzymes at a concentration of 1mg/ml can be used for preparation of protoplasts from Rhizoctonia solani.

Biochem/physiol Actions

Main enzymatic acitivity is β−(1-3)-glucanase, also reported to contain protease, pectinase, and amylase activities.

comparable product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of a cephalosporin C esterase gene in Acremonium chrysogenum for the direct production of deacetylcephalosporin C
Basch, J., et al
Journal of Industrial Microbiology, 31(11), 531-539 (2004)
D Tsuchiya et al.
Phytopathology, 91(4), 354-360 (2008-10-24)
ABSTRACT Cytological karyotypes with mitotic metaphase chromosomes were analyzed for Cochliobolus heterostrophus, C. carbonum, and C. sativus by the germ tube burst method (GTBM). Prior to karyotyping, procedures of GTBM suitable to Cochliobolus were established by examining several crucial conditions
Molecular characterization of a Stagonospora nodorum lipase gene LIP1
Feng, J., et al
Plant Pathology, 60(4), 698-708 (2011)
Structural characterization of a neutral antitumour β-D-glucan extracted with hot sodium hydroxide from cultured fruit bodies of Grifola frondosa.
Iino, K., et al
Carbohydrate Research, 141(1), 111-119 (1985)
Tung-Hsen Liu et al.
New biotechnology, 27(1), 64-69 (2009-09-01)
Novozym 234 was the most frequently used enzyme for production of Rhizoctonia solani protoplasts. Since manufacture of this enzyme was discontinued in the late 1990s, a new procedure was developed by testing lytic enzymes from Sigma and by examining factors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service