MilliporeSigma
Factors affecting protoplast formation by Rhizoctonia solani.
New biotechnology (2009-09-01)
Tung-Hsen Liu, Mei-Ju Lin, Wen-Hsiung Ko
ABSTRACT
Novozym 234 was the most frequently used enzyme for production of Rhizoctonia solani protoplasts. Since manufacture of this enzyme was discontinued in the late 1990s, a new procedure was developed by testing lytic enzymes from Sigma and by examining factors affecting protoplast formation. The combination of 20 mg/mL Driselase and 10mg/mL lysing enzyme was effective in releasing protoplasts from R. solani. The optimal condition for enzyme treatment of mycelium was incubation at 37 degrees C for 15 min followed by 34 degrees C for 105 min. The amount of protoplasts produced was positively correlated with growth rate and negatively correlated with mycelial density. Under favorable conditions, R. solani mycelia released 1.68 x 10(6) protoplasts/mL that is comparable with that produced with Novozym 234. Among various media tested, the best solid medium for protoplast regeneration was 1% V-8 juice agar, while the best liquid medium was 10% potato dextrose broth.
Sigma-Aldrich
Driselase Basidiomycetes sp., BioReagent, suitable for plant cell culture