Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5404000537

Eppendorf® Centrifuge 5424/5424R

5424R, refrigerated, with rotor FA-45-24-11, rotary knobs, AC/DC input 120 V, 60 Hz, US 3-pin plug (US)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701

feature

CSA approved
to hold 24 1.5/2.0 mL tubes
refrigerated
rotary knobs

manufacturer/tradename

Eppendorf® 5404000537 (NA)

parameter

-10-40 °C temp. range
100-15,000 rpm speed
21,130 × g max. RCF

technique(s)

centrifugation: suitable

W × H × D

29 cm × 48 cm × 26 cm

weight

21 kg

Looking for similar products? Visit Product Comparison Guide

General description

Centrifuge 5424 and Centrifuge 5424 R are the new laboratory standard. With their 24-place capacity and speed up to 21,130 × g they are perfectly equipped for all modern molecular biology applications in Eppendorf and PCR tubes. When working with spin columns kits, the Eppendorf Kit rotor with raised rim solves the issue of open tube caps shearing off during the elution step. Refrigerated Centrifuge 5424 R features state of the art cooling technology to ensure energy efficiency and high temperature accuracy for maximum sample protection. For ease-of-operation and time savings, Centrifuge 5424 R cools down to 4 deg. C in only 8 minutes.

Soft–touch, one–finger closure for ergonomic lid locking

Features and Benefits

  • Max. rotor capacity: 24 × 1.5/2.0 mL vessels
  • Max. speed: 20,238 × g (14,680 rpm)
  • OptiBowl® design for quiet operation, even without a rotor lid
  • Soft-touch one-finger lid closure for ergonomic operation
  • SOFT brake to protect sensitive samples
  • Full digital display with time and speed display
  • Separate short-spin key for quick centrifugation
  • Automatic lid opening at the end of the run – prevents sample warming and allows easy access to samples (only Centrifuge 5424)
  • Certified aerosol-tight rotor comes standard for safe centrifugation of hazardous samples
  • Available as a rotary knob version for fast parameter selection, or keypad version for easy cleaning


Special features for Centrifuge 5424 R
  • Temperature range from -10 to 40 deg. C
  • Guaranteed temperature of 4 deg. C at maximum rotational speed
  • FastTemp function for quick pre-cooling of the centrifuge, e.g. from approx. 21 to 4 deg. C in only 8 minutes
  • ECO power shut-off engages after 8 hours of non-use to reduce energy consumption (40 percent over night) and extend compressor life (can be deactivated)
  • The patented compressor technology reduces vibrations and protects your samples
  • Built-in condensation drain to eliminate water accumulation in the rotor chamber

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiBowl is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service