Skip to Content
MilliporeSigma
All Photos(1)

Documents

EP5406000445

Eppendorf® Centrifuge 5425 R

rotary knobs, refrigerated, AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug

Synonym(s):

Eppendorf® Centrifuge 5425

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701

feature

CE compliant
CSA approved
refrigerated
rotary knobs

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5406000445

parameter

21,300 × g speed

technique(s)

centrifugation: suitable

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The new 24–place Eppendorf Centrifuge 5425 R comes with an outstanding cooling system, 6 rotors, a max. speed of 21,300 × g and an ergonomic operation, making it the new favorite choice on the lab bench for the separation of temperature-sensitive samples from 0.2 – 5 mL.
Centrifuge 5425 is the new laboratory standard with its 24-place capacity and speed up to 21, 330 x G. Focusing on standard applications, it is an essential piece of equipment for modern labs ensuring optimal separation. The centrifuge 5425 serves as a reliable Partner especially for high-throughput users Expecting a top level of ergonomic comfort and provides great flexibility with its 6 rotor options.

Application

Centrifugation

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service