Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

EP022363344

Eppendorf® Safe-Lock microcentrifuge tubes

PCR clean, volume 2.0 mL, colorless, pack of 500

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
$135.00

$135.00


Available to ship onApril 24, 2025Details



Select a Size

Change View
500 EA
$135.00

About This Item

UNSPSC Code:
41121703

$135.00


Available to ship onApril 24, 2025Details


material

cap (safe-lock lid)
colorless
polypropylene

sterility

non-sterile

feature

PCR clean

packaging

pack of 500

manufacturer/tradename

Eppendorf® 022363344

parameter

30,000 × g max. RCF

technique(s)

PCR: suitable

volume

2.0 mL

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf® Safe-lock microtubes possess excellent centrifugation stability and well-optimized sealing of the tubes provides you with great confidence that your samples are in safe hands.

Application

Eppendorf® Safe-Lock microcentrifuge tubes are ideal for plasmid DNA recovery.

Features and Benefits

  • The hinged safe-lock lid avoids the unintentional opening of the lid while storing or incubating samples
  • Centrifugation stability up to 30,000 × g averts sample loss due to tube breakage and provides safety while using hazardous samples
  • Tubes contain no biocides, plasticizers, or slip agents
  • Minimal evaporation rates during sample storage due to precise lid sealing
  • Large, frosted lid and side surface for easy labeling
  • Excellent functionality between -86 °C to 100 °C
  • The tubes can be autoclaved when open at 121 °C for 20 mins
Tubes snap closed and stay closed. Flat top with writing surface for positive sample identification

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service