EP022363212
Eppendorf® Safe-Lock microcentrifuge tubes
size 1.5 mL, PCR clean, colorless, pkg of 500 ea
Synonym(s):
1.5 ml centrifuge tubes, polypropylene centrifuge tubes
Select a Size
$124.00
Select a Size
About This Item
$124.00
Recommended Products
material
polypropylene
sterility
non-sterile
feature
PCR clean
packaging
pkg of 500 ea
manufacturer/tradename
Eppendorf® 022363212
technique(s)
PCR: suitable
size
1.5 mL
color
colorless
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- Eppendorf® Safe-Lock microcentrifuge tubes are ideal for use in high demanding centrifugation and incubation applications
- High-quality polypropylene material offers excellent mechanical stability and chemical resistance
- Manufactured free from slip agents, plasticizers, and biocides that interfere with the samples
- The hinged lid ensures a leakproof seal and prevents accidental sample loss
- High centrifugation resistance of (up to 30,000 x g) prevents tube breakage
- Certified free from DNA, DNase, RNase, and PCR inhibitors
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service