Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU001091

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Elavl1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCTCAGAACATGACCCAAGAGGAACTACGAAGTCTGTTCAGCAGCATTGGCGAGGTTGAATCTGCAAAGCTTATTCGGGATAAAGTAGCAGGACACAGCTTGGGCTACGGTTTTGTGAACTATGTGACTGCAAAAGATGCAGAGAGAGCAATCAGCACACTGAACGGCTTGAGACTCCAGTCCAAAACCATTAAGGTGTCATATGCTCGCCCAAGCTCAGAGGTCATCAAAGATGCCAACTTATACATCAGTGGGCTCCCAAGGACCATGACACAGAAGGATGTGGAAGACATGTTTTCTCGGTTTGGGCGAATCATCAACTCCAGGGTCCTTGTGGATCAGACCACAGGTTTGTCCAGAGGGGTTGCCTTTATCCGGTTTGACAAACGGTCAGAAGCAG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danping Wu et al.
Clinical laboratory, 61(11), 1625-1634 (2016-01-07)
Increasing evidence suggests that microRNAs are widely involved in cancer progression and metastasis. However, the specific role of miR-31 in papillary thyroid carcinoma (PTC) is still largely unknown. The level of miR-31 and HuR was detected in 30 pairedcancerous and
Kotb Abdelmohsen et al.
Nucleic acids research, 42(15), 10099-10111 (2014-08-16)
Noncoding RNAs (ncRNAs) and RNA-binding proteins are potent post-transcriptional regulators of gene expression. The ncRNA 7SL is upregulated in cancer cells, but its impact upon the phenotype of cancer cells is unknown. Here, we present evidence that 7SL forms a
Kenneth K W To et al.
Experimental cell research, 338(2), 222-231 (2015-09-20)
Colorectal cancer (CRC) is a major cause of mortality and morbidity worldwide. While surgery remains the mainstay of treatment for early stage CRC, adjuvant chemotherapy is usually given to reduce the risk of recurrence after colectomy. Overexpression of a multidrug
Cristina M Ramírez et al.
Journal of lipid research, 55(6), 1066-1076 (2014-04-15)
ABCA1 is a major regulator of cellular cholesterol efflux and plasma HDL biogenesis. Even though the transcriptional activation of ABCA1 is well established, the posttranscriptional regulation of ABCA1 expression is poorly understood. Here, we investigate the potential contribution of the
Jeong-Dan Cha et al.
Head & neck, 36(8), 1168-1175 (2013-07-16)
HuR expression has been noted in several cancer types, in which it may contribute to increased expression of cellular inhibitors of apoptosis protein-2 (cIAP2) observed during tumorigenesis. To assess the correlation between cIAP2 and HuR in cases of oral squamous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service