Skip to Content
MilliporeSigma
All Photos(1)

Documents

V8380

Magenta vessel

GA-7-3

Synonym(s):

Magenta Vessel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

material

polycarbonate body
polypropylene closure

description

Autoclavable

W × L × H

77 mm × 77 mm × 77 mm

application(s)

agriculture

Looking for similar products? Visit Product Comparison Guide

General description

Unit is unbreakable and designed for prolonged use. Unique closure and drip ring makes this product ideal for plant tissue culture use.

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duan Chu et al.
BMC genetics, 21(1), 98-98 (2020-09-05)
C-to-U RNA editing is prevalent in the mitochondrial and chloroplast genes in plants. The biological functions of a fraction of C-to-U editing sites are continuously discovered by case studies. However, at genome-wide level, the cis and trans determinants affecting the
Alexandra K Nothstein et al.
PloS one, 14(4), e0214219-e0214219 (2019-04-20)
Selenium plays an important, but vastly neglected role in human nutrition with a narrow gap between dietary deficiency and toxicity. For a potential biofortification of food with Se, as well as for toxicity-risk assessment in sites contaminated by Se, modelling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service