MilliporeSigma
  • Cytoplasmic HuR expression: correlation with cellular inhibitors of apoptosis protein-2 expression and clinicopathologic factors in oral squamous cell carcinoma cells.
Cytoplasmic HuR expression: correlation with cellular inhibitors of apoptosis protein-2 expression and clinicopathologic factors in oral squamous cell carcinoma cells.
Head & neck (2013-07-16)
Jeong-Dan Cha, Hee-Kyeong Kim, In-Ho Cha
ABSTRACT
HuR expression has been noted in several cancer types, in which it may contribute to increased expression of cellular inhibitors of apoptosis protein-2 (cIAP2) observed during tumorigenesis. To assess the correlation between cIAP2 and HuR in cases of oral squamous cell carcinoma (OSCC), the expression patterns of HuR and cIAP2 were assessed by immunohistochemical analyses of 95 treated OSCC samples. In the tumor tissues, positive cytoplasmic HuR expression was evident in 71.6% of samples and positive cIAP2 expression was noted in 95.8% of samples. Positive cytoplasmic HuR expression was significantly associated with positive cIAP2 (p < .035) and high cIAP2 expression (p < .007), as well as high grade (p < .050). The inhibition of HuR expression by small interfering RNA or leptomycin B caused a reduction in the inducibility of cIAP2 in oral cancer cells. Cytoplasmic expression of HuR is associated with cIAP2 expression in OSCCs.
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Elavl1
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C