Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E9765

Sigma-Aldrich

Endothelial Cell Attachment Factor (ECAF)

liquid, sterile-filtered, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

sterility

sterile-filtered

Quality Level

form

liquid

packaging

pkg of 5 and 10 mL

storage condition

desiccated

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

shipped in

dry ice

storage temp.

−70°C

Application

ECAF is optimized to promote the attachment, spreading and polarity of endothelial cells. It contains gelatin (in HEPES buffered medium) to which factors separated from serum by chromatography and dialysis have been added. Use in combination with C1431, C1556, E9640 and T4299.

Formula variant

With gelatin, factors from iron-supplemented calf serum associated with endothelial cell attachment and spreading, sodium bicarbonate and HEPES buffers, and phenol red.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service