Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1556

Sigma-Aldrich

CS-C Medium Without Serum for Endothelial Cell Lines

With L-glutamine, sodium bicarbonate, and HEPES, without phenol red, liquid, sterile-filtered, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: no
HEPES: yes
L-glutamine: yes
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Application

CS-C Medium has been optimized for the culture of Human Umbilical Vein Endothelial Cells (HUVEC). It contains L-glutamine, sodium bicarbonate, and HEPES. It does not contain phenol red, serum or antibiotics.

Packaging

Packaged in PETG plastic bottles.

Reconstitution

Should be supplemented with 10% serum, Endothelial Cell Growth Factor (E 9640) and Endothelial Cell Attachment Factor (E 9765) for best results.

Other Notes

The selection of a nutrient medium is strongly influenced by 1] type of cell, 2] type of culture [monolayer, suspension, clonal] and 3] ° of chemical definition necessary. It is important to review the literature and more specific references for recommendations concerning medium, supplementation and physiological parameters required for a given cell.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Golam Sabbir et al.
Biochimica et biophysica acta. Molecular cell research, 1867(10), 118763-118763 (2020-06-03)
We recently reported that loss of calcium/calmodulin-dependent protein kinase kinase-2 (CAMKK2), a serine/threonine kinase activated by intracellular calcium, in mice leads to tissue-specific aberrant turnover of transferrin (TF), a receptor-mediated iron-transporter that supplies iron to tissues. Iron dyshomeostasis is associated
Jinhui Zhang et al.
Hearing research, 345, 10-22 (2017-01-15)
The vestibular blood-labyrinth barrier (BLB) is comprised of perivascular-resident macrophage-like melanocytes (PVM/Ms) and pericytes (PCs), in addition to endothelial cells (ECs) and basement membrane (BM), and bears strong resemblance to the cochlear BLB in the stria vascularis. Over the past

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service