Key Documents
CLS6791
Corning® LSE Benchtop Shaking Incubator with Platform
AC/DC input 230 V AC, Schuko plug
Synonym(s):
magnetic stirrer, magnetic stirrers, stirrers
About This Item
UNSPSC Code:
41104405
NACRES:
NB.73
Pricing and availability is not currently available.
feature
fits flask  4 x 1L
parameter
20-300 rpm shaking frequency
4-65 °C temp. range (Temperature Increments: 0.1 deg. C)
external W × D × H
37 cm × 53 cm × 40 cm
General description
Truly a benchtop style, the lid on the Corning benchtop shaking incubator has a low pivot point which provides a space-saving size sure to fit on the lab bench and under shelves. Each unit includes a software package, which provides the option of saving data to an external computer or monitoring rpm and temperature throughout runs. The maximum capacity is 4 x 1L flasks.
Features and Benefits
  • Outstanding temperature uniformity
  • Large viewing area for undisturbed monitoring of samples
  • Broad temperature range
  • Multiple shelving options
  • Precision control shaking mechanism
Legal Information
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)