Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST689

Ferrochromium silicon

NIST® SRM® 689

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$1,170.00

$1,170.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$1,170.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,170.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 100 g

manufacturer/tradename

NIST®

format

matrix material

General description

This SRM is in the form of fine powder for use in checking chemical methods of analysis andin calibration with instrumental methods of analysis. For more information, please refer to the SDS and COA.

SRM 689_cert

SRM 689_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Carbon (C), Manganese (Mn), Phosphorus (P), Sulfur (S), Silicon (Si), Copper (Cu), Nickel (Ni), Chromium (Cr), Vanadium (V), Aluminum (Al), Titanium (Ti), Cobalt (Co), Iron (Fe), Boron (B)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service