Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3251

Serenoa repens extract

NIST® SRM® 3251

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1 mL

manufacturer/tradename

NIST®

format

matrix material

General description

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Lipids and Fatty acids: Arachidic acid, Behenic acid, Behenic acid, Capric acid, Caproic acid, Caprylic acid, Caprylic acid, Erucic acid, Gondoic acid, Lauric acid, Lauric acid, Lignoceric acid, Lignoceric acid, Linoleic acid, Linoleic acid, Linolenic acid, Linolenic acid, Margaric acid, Margaric acid, Myristic acid, Myristic acid, Oleic acid, Oleic acid, Palmitic acid, Palmitic acid, Palmitoleic acid, Palmitoleic acid, Pentadecanoic acid (C15:0), Pentadecanoic acid (C15:0), Stearic acid, Stearic acid, Tridecanoic acid (C13:0), Tridecanoic acid (C13:0), Undecanoic acid (C11:0), Vaccenic acid, Vaccenic acid
Phytochemicals: Brassicasterol, Campesterol, Cycloartenol, Lupeol, Stigmasterol, β-Sitosterol
Vitamins: 9-cis-β-Carotene, Total β-carotene, trans-β-Carotene, γ-Tocopherol

Matrix Group: Botanicals

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Updated meta?analysis of clinical trials of Serenoa repens extract in the treatment of symptomatic benign prostatic hyperplasia.
Boyle P, et al.
BJU International, 93(6), 751-756 (2004)
Effect of the lipidic lipidosterolic extract of Serenoa repens ([TM="Permixon"]) on the ionophore A23187-stimulated production of leukotriene B4 (LTB4) from human polymorphonuclear neutrophils.
Paubert-Braquet M, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids, 57(3), 299-304 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service