Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2710A

Montana I soil

NIST® SRM® 2710a

Synonym(s):

Montana Soil Sample

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$1,880.00

$1,880.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$1,880.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,880.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for use in the analysis of soils, sediments, or other materials of a similar matrix. One unit of SRM 2710a consists of 50 g of the dried, powdered soil, blended with lead oxide. For more information, please refer to the COA and SDS.

SRM 2710A_cert

SRM 2710A_SDS

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Antimony (Sb), Arsenic (As), Barium (Ba), Calcium (Ca), Cadmium (Cd), Copper (Cu), Cobalt (Co), Iron (Fe), Lanthanum (La), Lead (Pb), Mercury (Hg), Magnesium (Mg), Strontium (Sr), Manganese (Mn), Uranium (U), Phosphorus (P), Potassium (K), Silicon (Si), Sodium (Na), Titanium (Ti), Zinc (Zn)

Reference/Informational value is provided for the following analytes.
Cesium (Cs), Chromium (Cr), Europium (Eu), Gadolinium (Gd), Lutetium (Lu), Neodymium (Nd), Nickel (Ni), Rubidium (Rb), Samarium (Sm), Scandium (Sc), Thallium (Tl), Thorium (Tr), Vanadium (V), Boron (B), Cerium (Ce), Dysprosium (Dy), Gold (Au), Hafnium (Hf), Indium (In), Selenium (Se), Silver (Ag), Tantalum (Ta), Terbium (Tb), Tungsten (W), Ytterbium (Yb), Zirconium (Zr)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Uncertainty contributions to species specific isotope dilution analysis. Part 1. Characterisation and stability of 199 Hg and 13 C isotopically enriched methylmercury by 1 H NMR.
Clough R, et al.
Journal of Analytical Atomic Spectrometry, 18(9), 1033-1038 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service