Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

67706

Millipore

Campylobacter Millichrome plus Supplement

25 L, selective for bacteria: Campylobacter spp., suitable for microbiology

Synonym(s):

Campylobacter Detection Supplement, Campylobacter Millichrome plus 25L Supplement, Millichrome Campylobacter plus Supplement, Supplement forCampylobacter Millichrome Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

sterility

sterile; 0.45 μm filtered

Quality Level

form

dry powder

usage

25 L

shelf life

limited shelf life, expiry date on the label

technique(s)

microbiological culture: suitable

solubility

sterile water: 21 mg/mL

application(s)

food and beverages
microbiology

shipped in

ambient

storage temp.

2-8°C

suitability

selective for bacteria: Campylobacter spp.

General description

Campylobacter Millichrome plus Supplement for 25 L is a selective supplement for the detection of thermotolerant Campylobacter and provides additional selectivity to the Campylobacter Millichrome plus Agar Base. The package contains 1 vial of supplement sufficient for 25 liters of final media. The supplement must be added to the melted Campylobacter Millichrome plus Agar Base at 45-50 °C.

Application

Campylobacter Millichrome plus Supplement for 25L is a selective supplement used in the qualitative direct detection, differentiation, and presumptive identification of thermotolerant Campylobacter. The supplement is used in the analyses of food products for human consumption, animal feed, and environmental samples.

Components

Composition:
(1 vial is sufficient for 25 L of final media)
Chromogenic and selective mix 5.25 g

Preparation Note

  • In a transparent vessel, add 210 mg of Campylobacter Millichrome plus Supplement to 10 ml of purified water.
  • Swirl well until complete dissolution.
  • Filter to sterilize at 0.45 μm.
  • Add the 10 ml of the supplement solution to 1 liter of the melted Campylobacter Millichrome plus Supplement for 5L at 45-50 °C.
  • Swirl or stir gently to homogenize.

Storage and Stability

  • Store in tightly sealed containers at 2-8°C.
  • Keep away from heat and sources of ignition.

Legal Information

MilliChrome is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service