Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

67683

Millipore

Campylobacter Culture Medium

MilliChrome, dry powder, final pH 7.3±0.2 (25 °C), suitable for microbiology

Synonym(s):

Campylobacter Millichrome plus Agar Base, Campylobacter chromogenic agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106204
NACRES:
NA.74

Agency

EP
JP
USP

Quality Level

Sterilization Compatibility

dry heat compatible

form

dry powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

MilliChrome

technique(s)

microbiological culture: suitable

final pH

7.3±0.2 (25 °C)

solubility

deionized water: 51.2 g/L

application(s)

food and beverages
microbiology

shipped in

ambient

storage temp.

15-25°C

suitability

selective for bacteria: Campylobacter spp.

General description

Campylobacter Millichrome plus Agar Base is a selective media for the detection of thermotolerant Campylobacter. The distinguishing feature of these media is the inclusion of a chromogenic substrate mix that produces visible color changes upon the enzymatic activity of Campylobacter.Millichrome chromogenic media is a specialized type of culture medium used to detect and isolate specific groups of bacteria based on their ability to produce enzymes with chromogenic substrates. Peptone and yeast extract provide nitrogenous nutrients for growth and other essential growth factors. Sodium chloride is needed for the osmotic balance. The chromogenic and selective mix contains chromogenic substrates for color differentiation based on the ability to cleave the substrate by characteristic enzymes. This mix contains as well selective agent that allows Campylobacter spp. to grow while other organisms are inhibited. Campylobacter Millichrome plus Supplement gives additional selectivity to this medium. Agar is the solidifying agent.

Application

Campylobacter Millichrome plus Agar Base is a selective chromogenic culture medium intended for use in the qualitative direct detection, differentiation and presumptive identification of pathogenic biotypes of thermotolerant Campylobacter from food, animal feed, and environmental samples

Preparation Note

Step 1 (Preparation of the base Campylobacter Millichrome plus Agar Base):

  • Disperse slowly 51.2 g of powder base in 1L of purified water.
  • Stir until agar is well thickened.
  • Heat and bring to boil (100 °C) while swirling or stirring regularly.
DO NOT HEAT TO MORE THAN 100 °C. DO NOT AUTOCLAVE AT 121 °C.
Warning: If using an autoclave, do so without pressure.
Advice: For the 100 °C heating step, mixture may also be brought to a boil in a microwave oven: after initial boiling, remove from oven, stir gently, then return to oven for short repeated bursts of heating until complete fusion of the agar grains has taken place (large bubbles replacing foam).
  • Cool in a water bath to 45-50 °C. Swirl or stir gently to homogenize.
Step 2 (preparation of the supplement):
  • In a transparent vessel, add 210 mg of Campylobacter Millichrome plus Agar Base Supplement in 10 ml of purified water.
  • Swirl well until complete dissolution.
  • Filter to sterilize at 0.45 μm.
  • Add the 10 ml of the supplement solution to the melted base (Step1) at 45-50 °C.
  • Swirl or stir gently to homogenize.
Step 3 (pouring):
  • Pour into sterile Petri dishes.
  • Let it solidify and dry (longer than usual).

Storage and Stability

Store prepared media below 8°C, protected from direct light (max. 1 month). Store dehydrated powder, in a dry place, in tightly sealed containers at 2-25°C.

Analysis Note

The Campylobacter colonies are colored red after 48 h of incubation at an optimal growth temperature of 42°C, whereas the growth of other species is inhibited or forms blue or green colonies after incubation at the same temperature.

Legal Information

MilliChrome is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service