Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

67432

Millipore

Campylobacter Millichrome plus Supplement

pkg of 5 L, suitable for microbiology

Synonym(s):

Campylobacter chromogenic Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

sterility

sterile; 0.45 μm filtered

Quality Level

form

dry powder

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 5 L

technique(s)

microbiological culture: suitable

solubility

sterile water: 21 mg/mL

application(s)

food and beverages
microbiology

shipped in

ambient

storage temp.

2-8°C

suitability

selective for bacteria: Campylobacter spp.

Related Categories

General description

Campylobacter Millichrome plus supplement is a selective media supplement for the detection of thermotolerant Campylobacter. The supplement provides additional selectivity to the Campylobacter Millichrome plus Agar Base. Millichrome chromogenic media is a specialized type of culture medium used to detect and isolate specific groups of bacteria based on their ability to produce enzymes with chromogenic substrates. The package contains 1 vial of supplement sufficient for 5 liters of final media. The supplement must be added to the melted Campylobacter Millichrome plus Agar Base at 45-50 °C.

Application

Campylobacter Millichrome plus supplement is a selective chromogenic culture media supplement intended for use in the qualitative direct detection, differentiation and presumptive identification of pathogenic biotypes of thermotolerant Campylobacter from various samples

Components

Composition in g/l:
(1 vial is sufficient for 5 L of final media)
Chromogenic and selective mix 1.05 g

Preparation Note

  • In a transparent vessel, add 210 mg of Campylobacter Millichrome plus Supplement to 10 ml purified water.
  • Swirl well until complete dissolution.
  • Filter to sterilize at 0.45 μm.
  • Add the 10 ml of the supplement solution to 1 liter of the melted Campylobacter Chromocult® plus Agar base at 45-50 °C.
  • Swirl or stir gently to homogenize.

Analysis Note

The Campylobacter colonies are colored red after 48 h of incubation at an optimal growth temperature of 42°C, whereas the growth of other species is inhibited or forms blue colonies after incubation at the same temperature.

Legal Information

CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
MilliChrome is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service