Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46122

Supelco

Ethylenediaminetetraacetic acid diammonium copper salt solution

volumetric, for ion-selective electrodes, 0.025 M EDTA-Cu(NH4)2

Synonym(s):

Copper diammonium EDTA, Diammonium copper ethylenediaminetetraacetate solution

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$124.00
1 L
$346.00

$124.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$124.00
1 L
$346.00

About This Item

Empirical Formula (Hill Notation):
C10H20CuN4O8
CAS Number:
Molecular Weight:
387.83
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

$124.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

for ion-selective electrodes

Quality Level

form

solid

quality

volumetric

shelf life

limited shelf life, expiry date on the label

concentration

0.025 M EDTA-Cu(NH4)2

SMILES string

[NH4+].[NH4+].[Cu++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.Cu.2H3N/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;2*1H3/q;+2;;/p-2

InChI key

QWQCKWBAUUOYMU-UHFFFAOYSA-L

General description

Visit our Sensor Applications portal to learn more.

Application

for complexometric titration with copper-selective electrode (Cu-ISE)

Caution

free of precipitate

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service