Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03668

Sigma-Aldrich

Ethylenediaminetetraacetic acid copper(II) disodium salt

≥97.0% (calculated on dry substance, RT)

Synonym(s):

(Ethylenedinitrilo)tetraacetic acid copper(II) disodium salt, Copper disodium ethylenediaminetetraacetate, EDTA copper(II) disodium salt, Edathamil

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$94.30
500 G
$378.00

$94.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$94.30
500 G
$378.00

About This Item

Linear Formula:
Cu(OOCCH2)2NCH2CH2N(CH2COONa)2
CAS Number:
Molecular Weight:
397.74
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$94.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (calculated on dry substance, RT)

form

powder

impurities

≤15% water

functional group

amine

SMILES string

[Na+].[Na+].[Cu++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.Cu.2Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;/q;+2;2*+1/p-4

InChI key

KCFCAUKZKOSSBI-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service