Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37205

Supelco

Nitrite test sticks Quantofix®

1-80 mg/L (NO2-)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

description

semi-quantitative

specific analyte(s)

nitrite

reaction suitability

reaction type: Griess reaction

measuring range

1-80 mg/L (NO2-)

application(s)

environmental
general analytical

detection method

colorimetric

General description

Contents:
Tube with 100 test sticks.

Storage:
Avoid exposing the strips to sunlight and moisture. Keep container cool and dry (store below +30 °C).

Gradation:
0 - 1 - 5 - 10 - 20 - 40 - 80 mg/l NO2-

Application

It is used to estimate the yield of the nitrate ion, in an experimental study of isoelectronic NO dioxygenase reaction, using a nonheme iron(III)-peroxo complex and nitrosonium ion.

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atsutoshi Yokoyama et al.
Chemical communications (Cambridge, England), 50(14), 1742-1744 (2014-01-08)
Reaction of a nonheme iron(III)-peroxo complex, [Fe(III)(14-TMC)(O2)](+), with NO(+), a transformation which is essentially isoelectronic with that for nitric oxide dioxygenases [Fe(III)(O2˙(-)) + NO], affords an iron(IV)-oxo complex, [Fe(IV)(14-TMC)(O)](2+), and nitrogen dioxide (NO2), followed by conversion to an iron(III)-nitrato complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service