Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.10057

Supelco

Nitrite Test Strips, colorimetric

greener alternative

0.5-10 mg/L (NO₂⁻), MQuant®

Synonym(s):

Nitrite chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Nitrite Test, colorimetric, 0.5-10 mg/L (NO2-), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

nitrite

reaction suitability

reaction type: Griess reaction

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

0.5-10 mg/L (NO2-)

detection method

colorimetric

greener alternative category

storage temp.

2-8°C

General description

Method: Colorimetric, with test strips, 0.5 - 1 - 2 - 5 - 10 mg/L (NO2-), MQuant®

The MQuant® The test can be used to determine nitrite in the environmental analysis, surface water investigations and aquaristics. The test extends the range of sensitivity into the lower range of detection. On the MQuant® Nitrite test strips, nitrite ions react with an aromatic amine to form a diazonium salt, which in turn reacts with N-(1-naphthyl)ethylenediamine to form a red-violet azo dye. The nitrite concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the nitrite concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® or Spectroquant® Nitrite test

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Application


  • Urinary nitrite/nitrate ratio measured by isotope-dilution LC-MS/MS as a tool to screen for urinary tract infections.: This study illustrates the application of the nitrite test in the rapid diagnosis of urinary tract infections using advanced liquid chromatography-mass spectrometry, enhancing the accuracy and speed of medical diagnostics (Chao et al., 2016).

  • Evaluations of urinary sediment and dry chemical detection in urinary tract infections.: This research evaluates the effectiveness of the nitrite test combined with dry chemical methods for diagnosing urinary tract infections, providing a comprehensive approach to enhance detection in clinical settings (Zhu et al., 2013).

  • New modification of the nitrite test in the diagnosis of urinary tract infections.: This historical study discusses a novel modification of the nitrite test aimed at improving the diagnostic process for urinary tract infections, demonstrating the evolution of analytical techniques in medical diagnostics (Sasinka et al., 1967).

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service