Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.10092

Supelco

Nitrate Test Strips, colorimetric

individually sealed, 10-500 mg/L (NO₃⁻), MQuant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Nitrate Test, colorimetric, 10-500 mg/L (NO3-), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

nitrate

reaction suitability

reaction type: Griess reaction

measuring range

10-500 mg/L (NO3-)

detection method

colorimetric

storage temp.

2-8°C

General description

Method: colorimetric with test strips (individually sealed ) 10 - 25 - 50 - 100 - 250 - 500 mg/l NO₃⁻ MQuant®
The nitrate test can be used for monitoring of drinking water, process water and wastewater, as well as aquarium water where limits can quickly be exceeded. It can also be used for determine nitrate in food e.g. potatoes.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service