Skip to Content
MilliporeSigma
Ethylenediaminetetraacetic acid copper(II) disodium salt ≥97.0% (calculated on dry substance, RT)

Ethylenediaminetetraacetic acid copper(II) disodium salt

Ethylenediaminetetraacetic acid copper(II) disodium salt ≥97.0% (calculated on dry substance, RT)
Synonym(s):
(Ethylenedinitrilo)tetraacetic acid copper(II) disodium salt, Copper disodium ethylenediaminetetraacetate, EDTA copper(II) disodium salt, Edathamil
Linear Formula:
Cu(OOCCH2)2NCH2CH2N(CH2COONa)2
CAS Number:
Molecular Weight:
397.74
EC Number:

Products

Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1