Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21-3280

Sigma-Aldrich

Nonne-Apelt′s reagent

Synonym(s):

Ammonium sulfate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
grade:
SAJ special grade
form:
liquid
Pricing and availability is not currently available.

grade

SAJ special grade

form

liquid

availability

available only in Japan

storage temp.

15-25°C

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Aqueous ammonium sulfate solution equivalent to indicated ammonia levels

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N A Nidialkova et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 74(5), 9-15 (2012-11-06)
Fibrinolytic peptidase of Bacillus thuringiensis IMV B-7324 was isolated by ammonium sulfate fractionation, gel-filtration and ion exchange chromatography on TSK-gels--Toyopearl HW-55 and DEAE 650 (M). Fibrinolytic activity of the purified enzyme was 87.9 U/mg of protein that was 19.9 times
Stefan Tenzer et al.
Methods in molecular biology (Clifton, N.J.), 960, 1-14 (2013-01-19)
Proteasomes are the main cytosolic proteases responsible for generating peptides for antigen processing and presentation in the MHC (major histocompatibility complex) class-I pathway. Purified 20S and 26S proteasomes have been widely used to study both specificity and efficiency of antigen
Soo Chan Lee et al.
Eukaryotic cell, 11(11), 1391-1398 (2012-09-25)
Cryptococcus neoformans is a human-pathogenic basidiomycete that commonly infects HIV/AIDS patients to cause meningoencephalitis (7, 19). C. neoformans grows as a budding yeast during vegetative growth or as hyphae during sexual reproduction. Pseudohyphal growth of C. neoformans has been observed
S Ishwarya et al.
Preparative biochemistry & biotechnology, 43(1), 95-107 (2012-12-12)
A serine protease was purified 6.9-fold from the leaves of Thespesia populnea using ammonium sulfate fractionation followed by CM-cellulose and Sephadex G-100 chromatography. The purified enzyme was named populnein and was characterized. It was made up of a single polypeptide
Urh Černigoj et al.
Journal of chromatography. A, 1281, 87-93 (2013-02-13)
To exploit different chromatographic modes for efficient plasmid DNA (pDNA) purification a novel monolithic chromatographic support bearing multimodal histamine (HISA) groups was developed and characterized. Electrostatic charge of HISA groups depends on the pH of the mobile phase, being neutral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service