MilliporeSigma
  • Purification of large cytosolic proteases for in vitro assays: 20S and 26S proteasomes.
Purification of large cytosolic proteases for in vitro assays: 20S and 26S proteasomes.
Methods in molecular biology (Clifton, N.J.) (2013-01-19)
Stefan Tenzer, Tobias Hain, Hendrik Berger, Hansjörg Schild
ABSTRACT
Proteasomes are the main cytosolic proteases responsible for generating peptides for antigen processing and presentation in the MHC (major histocompatibility complex) class-I pathway. Purified 20S and 26S proteasomes have been widely used to study both specificity and efficiency of antigen processing. Here, we describe the purification of active human 20S and 26S proteasomes from human erythrocytes by DEAE-ion exchange chromatography, ammonium sulfate precipitation, glycerol density gradient centrifugation, and Superose-6 size exclusion chromatography and their characterization using fluorogenic substrates and specific inhibitors.
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%