Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-5580

Sigma-Aldrich

Congo Red

JIS special grade

Synonym(s):

Congo Red 4B, Cosmos Red, Cotton Red B, Cotton Red C, Direct Red 28, Direct Red R, Direct Red Y

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H22N6Na2O6S2
CAS Number:
Molecular Weight:
696.66
Colour Index Number:
22120
Beilstein:
3894858
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

JIS special grade

availability

available only in Japan

mp

>360 °C (lit.)

SMILES string

[Na+].[Na+].Nc1c(cc(c2ccccc12)S([O-])(=O)=O)\N=N\c3ccc(cc3)-c4ccc(cc4)\N=N\c5cc(c6ccccc6c5N)S([O-])(=O)=O

InChI

1S/C32H24N6O6S2.2Na/c33-31-25-7-3-1-5-23(25)29(45(39,40)41)17-27(31)37-35-21-13-9-19(10-14-21)20-11-15-22(16-12-20)36-38-28-18-30(46(42,43)44)24-6-2-4-8-26(24)32(28)34;;/h1-18H,33-34H2,(H,39,40,41)(H,42,43,44);;/q;2*+1/p-2/b37-35+,38-36+;;

InChI key

IQFVPQOLBLOTPF-HKXUKFGYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianjun Qiao et al.
Orphanet journal of rare diseases, 7, 95-95 (2012-12-14)
Amyloidosis cutis dyschromica is a rarely documented variant of cutaneous amyloidosis. To date, only 26 cases have been reported. The purpose of this study was to improve the clinical and histopathological data for this variant of amyloidosis and to highlight
Thaís Dias Lemos Kaiser et al.
Diagnostic microbiology and infectious disease, 75(3), 235-239 (2013-01-15)
Staphylococcus epidermidis in immunocompromised patients can cause bacteremia related to the use of catheter due to biofilm production. There are different phenotypic methods to detect biofilm formation. One method is based on culture in brain heart infusion agar (BHIA) containing
Carmen Herrero de Dios et al.
Fungal genetics and biology : FG & B, 50, 21-32 (2012-11-15)
MAPK pathways are conserved and complex mechanisms of signaling in eukaryotic cells. These pathways mediate adaptation to different stress conditions by a core kinase cascade that perceives changes in the environment by different upstream elements and mediates adaptation through transcription
Julien Duboisset et al.
The journal of physical chemistry. B, 117(3), 784-788 (2013-01-08)
Amyloid fibrils are protein misfolding structures that involve a β-sheet structure and are associated with the pathologies of various neurodegenerative diseases. Here we show that Thioflavine-T and Congo Red, two major dyes used to image fibrils by fluorescence assays, can
Christina S Faherty et al.
PloS one, 7(11), e49980-e49980 (2012-11-21)
Shigella flexneri is a Gram-negative, facultative intracellular pathogen that causes millions of cases of watery or bloody diarrhea annually, resulting in significant global mortality. Watery diarrhea is thought to arise in the jejunum, and subsequent bloody diarrhea occurs as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service