MilliporeSigma
  • Modification of the Congo red agar method to detect biofilm production by Staphylococcus epidermidis.
Modification of the Congo red agar method to detect biofilm production by Staphylococcus epidermidis.
Diagnostic microbiology and infectious disease (2013-01-15)
Thaís Dias Lemos Kaiser, Eliezer Menezes Pereira, Kátia Regina Netto Dos Santos, Ethel Leonor Noia Maciel, Ricardo Pinto Schuenck, Ana Paula Ferreira Nunes
ABSTRACT
Staphylococcus epidermidis in immunocompromised patients can cause bacteremia related to the use of catheter due to biofilm production. There are different phenotypic methods to detect biofilm formation. One method is based on culture in brain heart infusion agar (BHIA) containing sucrose and red Congo dye (original Congo red agar). Our group created a new CRA formula and we have confirmed its capacity to detect biofilm production in 210 S. epidermidis strains, including 76 (36.2%) icaAB gene-positive strains. Other parameters were also evaluated. The new CRA formula that gave the best results was BHIA with sucrose (5%), Congo red (0.08%), NaCl (1.5%), glucose (2%), and vancomycin (0.5 mg/mL) (vancomycin-modified CRA-CRAmod). The CRAmod plus vancomycin may be a promising tool and can help to determine the real participation of S. epidermidis in the infectious process.
Product Description
Supelco
Congo Red, analytical standard
Sigma-Aldrich
Congo Red, certified by the Biological Stain Commission, BioXtra
Sigma-Aldrich
Congo Red, Dye content ≥35 %