Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01340

Supelco

Congo red (C.I. 22120)

pH indicator, Reag. Ph Eur

Synonym(s):

Congo red (C.I. 22120)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$135.00

$135.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$135.00

About This Item

Empirical Formula (Hill Notation):
C32H22N6Na2O6S2
CAS Number:
Molecular Weight:
696.66
MDL number:
UNSPSC Code:
12352107
EC Index Number:
209-358-4
NACRES:
NA.21

$135.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Congo red (C.I. 22120), indicator Reag. Ph Eur

Agency

reag. Ph. Eur.

Quality Level

vapor pressure

<1 Pa

form

powder

potency

143 mg/kg LD50, oral (Rat)

loss

≤5% loss on drying, 110°C

color

almost clear to slightly turbid red, 1.0g/L;water
red-brown

pH

6.7 (20 °C, 10 g/L in H2O)

mp

>360 °C

bulk density

600‑700 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])c1c2c(c(c(c1)N=Nc3ccc(cc3)c4ccc(cc4)N=Nc5c(c6c(c(c5)[S](=O)(=O)[O-])cccc6)N)N)cccc2

InChI

1S/C32H24N6O6S2.2Na/c33-31-25-7-3-1-5-23(25)29(45(39,40)41)17-27(31)37-35-21-13-9-19(10-14-21)20-11-15-22(16-12-20)36-38-28-18-30(46(42,43)44)24-6-2-4-8-26(24)32(28)34;;/h1-18H,33-34H2,(H,39,40,41)(H,42,43,44);;/q;2*+1/p-2

InChI key

IQFVPQOLBLOTPF-UHFFFAOYSA-L

Analysis Note

Appearance: Red-brown to brownishred powder, eventually with granular parts.
Appearance of solution (1.0 gr/l; water): Almost clear to slightly turbid, red
Transition range: pH 3.0 - pH 5.2violet - orange-red
Loss on drying (110 °C): ≤ 5 %
Sensitivity test: passes test

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service