Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX7132

Sigma-Aldrich

Anti-hu N-term proBNP (NT-proBNP) clone 1312

clone 13G12, from mouse

Synonym(s):

proBNP, Brain neurotrophic peptide, NT-proBNP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

13G12, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2a

Gene Information

human ... NPPB(4879)

General description

Antibody reacts with the pro-form of Brain Neurotrophic Peptide (NT-proBNP). BNP is a protein that is released into the blood due to excessive stretching of the heart muscle cells. proBNP is a marker that is routinely used in the diagnosis of specific cardiac events

Immunogen

proBNP

Application

Confirmed matched pairing. Capture Antibody MABX7132-1KC; Detector Antibody MABX7154-1KC
ELISA;Western Blotting
Research Category
Cancer

Physical form

Format: Purified
PBS (pH 7.4) with 0.09% sodium azide
Protein A purified

Storage and Stability

Stable for 12 months at 2-8ºC from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service