Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABS2307

Sigma-Aldrich

Anti-JMY Antibody, clone HMY 117

Synonym(s):

Junction-mediating and -regulatory protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

HMY 117, monoclonal

mol wt

calculated mol wt 111.45 kDa
observed mol wt ~130 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... JMY(133746)

Related Categories

General description

Junction-mediating and -regulatory protein (UniProt: Q8N9B5; also known as JMY) is encoded by the JMY gene (Gene ID: 133746) in human. JMY is involved in a complex pathway connecting cell motility and invasion with the p53 response to DNA damage. It is widely expressed in many types of tissue- with cytoplasmic and nuclear co-expression. It contains a unique N-terminal domain (aa 1-126) that is essential for its interaction with p300/EP300; three central coiled-coil segments (aa 324-360; 489-541; and 590-621), a polyproline region (aa 801-830), and a C-terminal WH2 domain (aa 921-938). It localizes primarily to the nucleus, but is also found at the leading edge in migrating cells. It acts both as a nuclear p53/TP53-cofactor and a cytoplasmic regulator of actin dynamics depending on conditions. In the nucleus, it acts as a cofactor that increases p53/TP53 response via its interaction with p300/EP300 and increases p53/TP53-dependent transcription and apoptosis. In the cytoplasm, it acts as a nucleation-promoting factor for both branched and unbranched actin filaments. It promotes the rapid formation of a branched actin network by first nucleating new mother filaments and then activating Arp2/3 to branch off these filaments. However, it does not affect elongation of preformed filaments. JMY is shown to be expressed in every type of tumor investigated. Overexpression of JMY in human U2OS cells is shown to induce the formation of elongated actin filament structures. JMY is ubiquitinated by MDM2 that leads to its proteasomal degradation. In case of DNA damage its interaction with MDM2 is altered that prevents its degradation and allows interaction with p300/EP300. (Ref.: Adighibe, O., et al. (2014). Virchows Arch. 465(6); 715-722; Firat-Karalar, EN., et al. (2011). Mol. Biol. Cell. 22(23); 4563-4574; Zuchero, JB., et al. (2009). Nat. Cell Biol. 11(4); 451-459).

Specificity

Clone HMY 117 is a mouse monoclonal antibody that detects Junction-mediating and -regulatory protein (JMY).

Immunogen

KLH-conjugated three peptides corresponding to 13 amino acids from the N-terminal region, 14 amino acids from the internal region, and 23 amino acids from the C-terminal region of human Junction-mediating and -regulatory protein (JMY).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Junction-mediating and -regulatory protein (JMY) in HeLa cell lysates.

Tested Applications

Immunohistochemistry Applications: A representative lot detected JMY in Immunohistochemistry applications (Adighibe, O., et al. (2014). Virchows Arch. 465(6):715-22).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-JMY, clone HMY 117, Cat. No. MABS2307, is a mouse monoclonal antibody that detects Junction-mediating and -regulatory protein (JMY) and is tested for use in Immunohistochemistry and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service