Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCM082

Sigma-Aldrich

ES2N Complete Medium Kit

The ES2N Complete medium is a defined serum-free formulation that has been optimized for the differentiation of mouse embryonic stem (ES) & induced pluripotent stem (iPS) cells into neurons.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.71
Pricing and availability is not currently available.

Quality Level

form

liquid

technique(s)

cell culture | stem cell: suitable

input

sample type induced pluripotent stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type neural stem cell(s)

shipped in

dry ice

General description

ES2N Complete medium is a defined serum-free formulation that has been optimized for the differentiation of mouse embryonic stem (ES) and induced pluripotent stem (iPS) cells into neurons. When used in conjucture with ESGRO Complete Plus medium mouse ES and iPS cells readily differentiate into neurons in a monolayer assay within 9-12 days on gelatin coated culture dishes. When supplemented with bFGF and EGF, this medium enables the isolation and expansion of neural stem cells (NSCs).

Application

The ES2N Complete medium is a defined serum-free formulation that has been optimized for the differentiation of mouse embryonic stem (ES) & induced pluripotent stem (iPS) cells into neurons.

Components

ES2N Basal Medium: (Cat. No. SCM083) One (1) 245 mL bottle containing standard base medium.

Neuro27 Medium Supplement: (Cat. No. SCM013-S) One (1) 5 mL vial containing neural supplement (use as 50x).

Neuro2 Medium Supplement: (Cat. No. SCM012-S) One (1) 1.25 mL vial containing N2-like supplement (use as 200x).

Quality

Cell Proliferation Assay: The biological activity of this lot of VEGF was determined by its mitogenic activity on human umbilical vein endothelial cells (HUVEC) with an ED50 of about 1.0 ng/mL. Results may vary depending on the cell line used. VEGF will also stimulate endothelial cells from other species.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service