Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2037

Sigma-Aldrich

Anti-Factor XII Antibody, clone 15H8

clone 15H8, from mouse

Synonym(s):

Coagulation factor XII, EC: 3.4.21.38, Hageman factor, HAF

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00
100 μG
$607.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$231.00
100 μG
$607.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15H8, monoclonal

species reactivity

human, baboon

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... F12(2161)

General description

Coagulation factor XII (UniProt: P00748; also known as EC: 3.4.21.38, Hageman factor, HAF) is encoded by the F12 gene (Gene ID: 2161) in human. Factor XII is a serum glycoprotein that participates in the initiation of blood coagulation, fibrinolysis, and the generation of bradykinin and angiotensin. It activates factor XI to XIa and factor VII to VIIa by selective cleavage of Arginine-Isoleucine bond. Factor XII itself can be activated to factor XIIa either by exposure to negatively charged surfaces or via proteolytic cleavage. It is synthesized in the liver with a signal peptide (aa 1-19), which is subsequently cleaved off in the mature form. Coagulation factor XII circulates in blood as a single chain zymogen, which is converted to a two-chain serine protease with a heavy chain (alpha-factor XIIa) and a light chain. The heavy chain contains two fibronectin-type domains, two epidermal growth factor (EGF)-like domains, a kringle domain, and a proline-rich domain. The light chain contains only a catalytic domain. Mutations in F12 gene can lead to Factor XII deficiency that is generally an asymptomatic anomaly of in vitro blood coagulation. However, some mutations may cause hereditary angioedema 3, which is characterized by episodic local swelling involving subcutaneous or submucous tissue of the upper respiratory and gastrointestinal tracts, face, extremities, and genitalia.

Specificity

Clone 15H8 is a mouse monoclonal antibody that specifically detects Coagulation factor XII.

Immunogen

A mixture of purified human Factor XII and Factor XIIa

Application

Anti-Factor XII, clone 15H8, Cat. No. MABS2037, is a mouse monoclonal antibody that detects Coagulation factor XII and has been tested for use in ELISA, Inhibition and Function assays, and Western Blotting.
Function Analysis: A representative lot prolonged the activated partial thromboplastin time (aPTT). (Matafonov, A., et. al. (2014). Blood. 123(11):1739-46).

Inhibition Analysis: A representative lot inhibited the generation of thrombin in normal plasma following addition of activated partial thromboplastin reagent. (Matafonov, A., et. al. (2014). Blood. 123(11):1739-46).

Western Blotting Analysis: A representative lot detected Factor XII in Western Blotting applications (Matafonov, A., et. al. (2014). Blood. 123(11):1739-46).

ELISA Analysis: A representative lot detected Factor XII in ELISA applications (Matafonov, A., et. al. (2014). Blood. 123(11):1739-46).
Research Category
Signaling

Quality

Evaluated by Western Blotting in human factor XII.

Western Blotting Analysis: 1 µg/mL of this antibody detected human Coagulation factor XII.

Target description

~75 kDa observed; 67.79 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in PBS, pH 7.4, without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service