Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2156

Sigma-Aldrich

Anti-Thrombomodulin Antibody, clone 1009

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$242.00
100 μL
$619.00

$242.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$242.00
100 μL
$619.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$242.00


Please contact Customer Service for Availability

Request a Bulk Order

antibody form

purified antibody

clone

1009
monoclonal

purified by

using protein G

species reactivity

human, rat, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

Mode of action

| inhibits

General description

Recombinant fragment corresponding to 241 amino acids corresponding to six EGF repeats from human Thrombomodulin.

Specificity

Clone 1009 is a mouse monoclonal antibody that detects Thrombomodulin.

Application

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Thrombomodulin in human tonsil tissue sections.

ELISA Analysis: A representative lot detected Thrombomodulin in ELISA applicaitons (Woodley-Cook, J., et. al. (2006). Mol Cancer Ther. 5(12):3303-11).

Radioimmunoassay Analysis: A representative lot detected Thrombomodulin in Radioimmunoassay applications (Woodley-Cook, J., et. al. (2006). Mol Cancer Ther. 5(12):3303-11).

Inhibits Activity/Function Analysis: A representative lot of this blocked thrombin binding and reduced Protein C activation (Stearns-Kurosawa, D.J., et. al. (1996). Proc Natl Acad Sci USA. 93(19):10212-6).

Flow Cytometry Analysis: A representative lot detected Thrombomodulin in Flow Cytometry applications (Stearns-Kurosawa, D.J., et. al. (1996). Proc Natl Acad Sci USA. 93(19):10212-6; Woodley-Cook, J., et. al. (2006). Mol Cancer Ther. 5(12):3303-11).

Western Blotting Analysis: A representative lot detected Thrombomodulin in Western Blotting applications. (Xu, J., et. al. (1999). J Biol Chem. 274(10):6704-10; Woodley-Cook, J., et. al. (2006). Mol Cancer Ther. 5(12):3303-11).

Immunocytochemistry Analysis: A representative lot detected Thrombomodulin in Immunocytochemistry applications (Campbell, J.E., et. al. (2010). Blood. 116(26):6082-91).

Immunohistochemistry (Paraffin) Analysis: A representative lot detected Thrombomodulin in Immunohistochemistry applications (Trotman, W.E., et. al. (2011). Histochem Cell Biol. 135(2):141-52).

Quality

Evaluated by Immunohistochemistry (Paraffin) in human placenta tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected Thrombomodulin in human placenta tissue sections.

Physical properties

~60.33 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Human, Rat.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service