Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABC950

Sigma-Aldrich

Anti-Aminopeptidase N/CD13 Antibody, clone SL13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

Quality Level

clone

SL13
monoclonal

purified by

using protein G

species reactivity

mouse, rat

concentration

(Please refer to lot specific datasheet.)

technique(s)

activity assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

UniProt accession no.

General description

Recombinant protein corresponding to mouse Aminopeptidase N/CD13.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Aminopeptidase N/CD13 in mouse kidney cortex, mouse kidney medulla, and mouse colon tissue.
Immunohistochemistry Analysis: A representative lot detected myocardial infarction-/MI-induced infiltration of aminopeptidase N/CD13-positive dendritic cells in murine cardiac tissue (Pereira, F.E., et al. (2013) Cardiovasc. Res. 100(1):74-83).
Western Blotting Analysis: A representative lot detected aminopeptidase N/CD13 upregulation in murine cardiac tissue following myocardial infarction (MI) induction (Pereira, F.E., et al. (2013) Cardiovasc. Res. 100(1):74-83).
Activity Assay Analysis: A representative lot observed Aminopeptidase N/CD13 blocking monocyte trafficking (Subramani, J., et al. (2014). J. Immunol. 191(7):3905-3912).
Activity Assay Analysis: A representative lot observed Aminopeptidase N/CD13 blocks the adhesion to matrigel of WT mouse satellite cells (Rahman. M.M., et al. (2014). Stem Cells. 32(6):1564-1577).
Western Blotting Analysis: A representative lot detected Aminopeptidase N/CD13 in primary muscle satellite cells (Rahman. M.M., et al. (2014). Stem Cells. 32(6):1564-1577).

Quality

Evaluated by Western Blotting in mouse kidney tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Aminopeptidase N/CD13 in 10 µg of mouse kidney tissue lysate.

Physical properties

~160 kDa observed. The calculated molecular weight is 110 kDa, however Aminopeptidase N/CD13 has been shown as a ~160 kDa band in western blots (Soderberg, et al. (1993). Journal of virology).

Physical form

Purified rat monoclonal IgG2bκ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service