Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

164605

Sigma-Aldrich

Aminopeptidase N (CD13), Human Serum

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$543.00

$543.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$543.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

$543.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Storage Class Code

10 - Combustible liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service