Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

164606

Sigma-Aldrich

Aminopeptidase N/CD13 Inhibitor

The Aminopeptidase N/CD13 Inhibitor controls the biological activity of Aminopeptidase N/CD13. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

Aminopeptidase N/CD13 Inhibitor, 4-tert-butylphenyl hydrogen 1-aminopropylphosphonate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$181.00

$181.00


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
$181.00

About This Item

Empirical Formula (Hill Notation):
C13H22NO3P
Molecular Weight:
271.29
UNSPSC Code:
12352200
NACRES:
NA.77

$181.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

0.1 M NaOH: 1 mg/mL
DMSO: 1 mg/mL (Only partially soluble)
water: 1 mg/mL (only partially soluble)

shipped in

wet ice

storage temp.

−20°C

General description

An alpha-aminoaralkylphosphonic acid ester that acts as an aminopeptidase N/CD13 inhibitor (IC50=1.1 µM). Cell growth is dose dependently inhibited by as much as ~36% and ~60% in PANC-1 and HT-1080 cells, respectively when 300 µM of compound 4d is co-treated with 30 µg/ml of TNF. When treated with compound 4d alone, cell growth is dose-dependently inhibited by as much as 26% and 36%, in the same cell lines, respectively.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Grzywa, R., et al. 2010. Bioorg. Med. Chem.8, 2930.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service