Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EZSH0SBB2

Millipore

Hoses barbs for EZ-Stream® pump

To replace IN and OUT original hose barbs

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$94.10

$94.10


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EACH
$94.10

About This Item

UNSPSC Code:
41116500
NACRES:
NB.79

$94.10


Please contact Customer Service for Availability

Request more information

description

hose barb adapter

feature

repair kit

packaging

pkg of 2 numbers

manufacturer/tradename

EZ-Stream®

application(s)

bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring

compatibility

for use with EZ-Stream®

General description

The hose barbs are for replacement of original parts if they are broken or in case of a leak in the EZ-Stream® vacuum pump and ensures proper tubing connection. The EZ-Stream® vacuum pump is a liquid transfer pump with a compact design that makes the pump ideal for use on both the workbench and in the laminar flow hood. Two hoses barbs and two O-rings are included in order to replace parts connected to inlet and outlet tubing.

Application

The hose barbs ensure proper tubing connection of the EZ-Stream® pump that is used for microbiology analysis of Beer, Dairy, Bottled Water, Pharmaceutical Products Analysis, Soft Drink, Cider, Sports Drink & Wine.

Components

  • 2 Hose barbs
  • 2 O-Rings

Legal Information

EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service