Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EZSKTMEM1

Millipore

Liquid pump service kit for EZ-Stream® pump

For servicing of the liquid pump of the EZ-Stream®

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$941.00

$941.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EACH
$941.00

About This Item

UNSPSC Code:
41116500
NACRES:
NB.79

$941.00


Please contact Customer Service for Availability

Request more information

feature

repair kit

manufacturer/tradename

EZ-Stream®

application(s)

bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring

compatibility

for use with EZ-Stream®

General description

The maintenance kit is a complete set of elements required to service the liquid pump of the EZ-Stream® and ensure optimal performance. The EZ-Stream® vacuum pump is a liquid transfer pump with a compact design that makes the pump ideal for use on both the workbench and in the laminar flow hood. The maintenance kit includes wear parts that can be replaced inside the liquid pump to keep high performance flow rate.

Application

The maintenance kit for the liquid pump of the EZ-Stream® ensures optimal performance of the pump that is used for microbiology analysis of Beer, Dairy, Bottled Water, Pharmaceutical Products Analysis, Soft Drink, Cider, Sports Drink & Wine.

Components

  • 2 flapper valves
  • 1 working membrane
  • 2 O-rings
  • 1 oscillation membrane

Legal Information

EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service