Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SYMBLFH01

Millipore

Steritest® Symbio LFH Pump Kit

The Steritest Symbio LFH Pump, designed to fit your testing environment

Synonym(s):

Peristaltic pump, Sterility Testing pump

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$28,980.00
1 EACH
$31,070.00

$28,980.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
$28,980.00
1 EACH
$31,070.00

About This Item

UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
NB.75

$28,980.00


Please contact Customer Service for Availability

Request more information

AC/DC input

100 V / 240 V AC, 50-60 Hz

Quality Level

material

316L stainless steel cover (for pump head)
316L stainless steel head (of pump)
316L stainless steel holder (for bottle)
316L stainless steel housing (for screen)
316L stainless steel pump
316L stainless steel support (for drain tray)
EPDM rubber seal
glass window(s) (toughened)
polyester keys
polyphenylsulfone (PPSU) container (for drain tray)
polyphenylsulfone (PPSU) guide(s) (for tubing)
silicon seal

Agency

EP 2.6.1
JP 4.06
USP 71

packaging

pkg of 1 unit

manufacturer/tradename

Steritest®

H × W × depth

41.0 cm (16.1 in.) × 41.0 cm (16.1 in.) × 37.2 cm (14.6 in.), with accessories

pump body H × W × depth

27.2 cm (10.7 in.) × 33.3 cm (13.1 in.) × 37.2 cm (13.1 in.)

pump head H

15.8 cm (6.2 in.)

weight

14.2 kg (31.3 lb ), (pump body)
15.8 kg (34.8 lb)with standard accessories

input

sample type pharmaceutical(s)

application(s)

pharmaceutical
sterility testing
(Industrial microbiology)

compatibility

standard for VHP

General description

The Steritest® Symbio LFH Pump with its compact design can be used comfortably in smallest testing environments. In the standard configuration this pump is used, in a laminar flow hood, a biosafety cabinet, or in a clean room. However, it can also be used in an isolator, using Steritest® Symbio high feet and the Steritest® Symbio connection cable extension.
The automatic closure of the pump head eliminates the risk of glove pinching and ensures reliable splitting of the liquid sample. As ergonomic working conditions are key, the drain tray and bottle holder can be easily and flexibly positioned around the pump head for easy access in limited-space environments. Furthermore, the drain tray is designed to minimize splashing under the canister base. The pump′s convenient pressure detection system which can be run in two different modes (pressure monitoring and pressure regulation), reduces the risk of false test result and sterility test interruption. For highest precision and reproducible sampling also of smallest volumes, the pump has a convenient timer function available. To further ensure test reproducibility and to be always compliant with the defined test procedures, the respective test methods can be created on a PC using the Steritest® Symbio Software. These test methods will then be transferred to the pump and will lead the operator through the workflow step-by-step.
A wide range of optional accessories are also offered to improve the sterility testing workflow.

Application

Steritest® Symbio LFH Pump (2 media) is suitable for sterility testing in the laminar flow hood, biosafety cabinet, cleanroom or inside an isolator. The sterile products that can be tested with the Steritest® system include, large- and small-volume parenteral solutions in glass or plastic bottles, collapsible bags, ampoules, and vials, pre-filled syringes, lyophilized and other soluble products in ampoules or vials, antibiotics in ampoules or vials, medical devices, and difficult-to-dissolve powders.

Features and Benefits

  • Easy-to-use design
  • Reliable
  • Safe
  • Ergonomic
  • User-friendly

Components

Peristaltic pump

Guides quality certificate

Bottle holder

Drain tray

Silicone drain tubing

Communication hub

Power cord

Network cable

USB flash drive

3 quick guides

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

accessory

Product No.
Description
Pricing

consumable

Product No.
Description
Pricing

recommended

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service