Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AG209

Sigma-Aldrich

VGT 2 Peptide

control for AB5907, liquid

Synonym(s):

VGLUT2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

product name

Vesicular Glutamate Transporter 2, control peptide for AB5907,

biological source

rat

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

cell based assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

General description

Synthetic peptide from rat VGLUT2 protein.
Vesicular Glutamate Transporter 2 (VGLUT2) is a sodium dependent inorganic phosphate cotransporter that is involved in the calcium dependent glutamate release from astrocytes. It is expressed in only a subset of neurons, localized to synaptic vesicles, at synapses exhibiting classical excitatory features. VGLUT2 mRNA is found in brain regions that lack VGLUT1, a related protein also displaying sodium dependent inorganic phosphate cotransporter activity.

Application

For use in blocking the reactivity of AB5907.

Optimal working dilution must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Neuronal & Glial Markers

Physical form

Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 3 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuroanatomical plasticity in the gonadotropin-releasing hormone system of the ewe: seasonal variation in glutamatergic and gamma-aminobutyric acidergic afferents.
Anna Sergeeva,Heiko T Jansen
The Journal of Comparative Neurology null
Sprouting of colonic afferent central terminals and increased spinal mitogen-activated protein kinase expression in a mouse model of chronic visceral hypersensitivity.
Andrea M Harrington,Stuart M Brierley,Nicole Isaacs,Patrick A Hughes,Joel Castro,L Ashley Blackshaw
The Journal of Comparative Neurology null
Marie-Madeleine Gabellec et al.
The European journal of neuroscience, 25(5), 1373-1383 (2007-04-12)
Vesicular glutamate transporters (VGLUTs) mediate the packaging of the excitatory neurotransmitter glutamate into synaptic vesicles. Three VGLUT subtypes have so far been identified, with distinct expression patterns in the adult brain. Here, we investigated the spatial distribution of the three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service