Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71251-M

Sigma-Aldrich

Veggie NovaBlue Singles Competent Cells - Novagen

Escherichia coli, rod shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202

product name

Veggie NovaBlue Singles Competent Cells - Novagen,

biological source

Escherichia coli

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: endA1 hsdR17 (rK12 mK12+) supE44 thi-1 recA1 gyrA96 relA1 lac F′[proA+B+ lacIqZΔM15::Tn10] (TetR)





This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Veggie NovaBlue Singles Competent Cells are available for routine cloning applications. NovaBlue is a K-12 strain ideally suited as an initial cloning host due to its high transformation efficiency, blue/white screening capability (with appropriate plasmids), and mutations in recA and endA, which result in high yields of high-quality plasmid DNA.

Features and Benefits

• Guaranteed efficiency >1.5 × 10⁸ cfu/µg

• Manufactured free of animal-derived mediaand components

• Reproducible High-efficiency cloning

• Produces high-quality plasmid DNA

• Easy-to-use Singles format

• Prepared using an optimized chemical method

Components

•11 or 22 rxnVeggie NovaBlue Singles Competent Cells

•2 or 4 × 2 mlSOC Medium

•10 µlTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

DNA Profile

Genotype: endA1 hsdR17 (rK12 mK12+) supE44 thi-1 recA1 gyrA96 relA1 lac Fʹ[proA+B+ lacIqZΔM15::Tn10] (TetR)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service